missing translation for 'onlineSavingsMsg'
Learn More

Cadmium foil, 1.0mm (0.04in) thick, Puratronic™, 99.998% (metals basis)

Product Code. 11371149
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11371149

Brand: Thermo Scientific Alfa Aesar 011368.GH

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-43-9 | Cd | 112.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium(2+) ion
SMILES [Cd]

Specifications

Quantity 100 x 100 mm
Weight ≈21.6g/50x50mm
Form Foil
Percent Purity 99.998%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Cadmium foil, 1.0mm (0.04 in.) thick

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.