missing translation for 'onlineSavingsMsg'
Learn More

Cadmium hydroxide, Puratronic™, 99.98% (metals basis)

Product Code. 11320889
Click to view available options
Quantity:
2 g
10 g
25 g
Unit Size:
10g
25g
2g
This item is not returnable. View return policy

Product Code. 11320889

Brand: Thermo Scientific Alfa Aesar 010662.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21041-95-2 | CdH2O2 | 146.43 g/mol

It is found in the industrial nickel cadmium storage batteries, in cadmium plating, in the manufacture of cadmium salts, laboratory reagents. It is a useful biochemical for proteomics research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21041-95-2
Molecular Formula CdH2O2
Molecular Weight (g/mol) 146.43
MDL Number MFCD00015995
InChI Key PLLZRTNVEXYBNA-UHFFFAOYSA-L
IUPAC Name cadmium(2+) dihydroxide
SMILES [OH-].[OH-].[Cd++]

Specifications

Melting Point 300°C (decomposition)
Physical Form Powder
Quantity 2 g
UN Number UN2570
Merck Index 14,1620
Solubility Information Soluble in dilute acids. Slightly soluble in water; 0.026g/100 mL.
Formula Weight 146.41
Percent Purity 99.98%
Grade Puratronic
Sensitivity Air Sensitive
Density 4.79 g/mL at 15°C
Assay (metals basis)
Chemical Name or Material Cadmium hydroxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.