missing translation for 'onlineSavingsMsg'
Learn More

Cadmium ingot/button,≈36mm (1.4 in.) dia. x 8mm (0.3 in.) thick, 99.9% (metals basis), Thermo Scientific™

Product Code. 15444945
Click to view available options
Quantity:
1 Pc.
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Color Blue-White
CAS 7440-43-9
Molecular Formula Cd
MDL Number MFCD00010914
Quantity 1 Pc.
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES [Cd]
IUPAC Name cadmium
Molecular Weight (g/mol) 112.41
PubChem CID 23973
ChEBI CHEBI:22977
Percent Purity 99.9%
Weight ≈50g/piece
Form Pieces
Assay (metals basis)
Chemical Name or Material Cadmium ingot/button, ≈36mm (1.4 in.) dia. x 8mm (0.3 in.) thick
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'