missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium iodide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.
Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It finds it use in photography, process engraving, lithography, analytical chemistry, and electroplating; Also used as in lubricants, phosphors. Used for cadmium electrode position and in photoconductors.
Solubility
Very soluble in water, slightly soluble in acetone, ethanol and diethyl ether.
Notes
Light Sensitive & Hygroscopic. Store away from oxidizing agents, light and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
Boiling Point | 787°C |
Melting Point | 387°C to 389°C |
Quantity | 25 g |
UN Number | UN2570 |
Merck Index | 14,1621 |
Solubility Information | Very soluble in water,slightly soluble in acetone,ethanol and diethyl ether. |
Formula Weight | 366.21 |
Percent Purity | 98% |
Odor | Odorless |
Sensitivity | Hygroscopic; Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'