missing translation for 'onlineSavingsMsg'
Learn More
Cadmium nitrate tetrahydrate, 98.5% min, Thermo Scientific Chemicals Product Code.: 11346648

Cadmium nitrate tetrahydrate, 98.5% min, Thermo Scientific Chemicals

Product Code. 11346648
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11346648

Brand: Thermo Scientific Alfa Aesar 012228.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium nitrate is used in manufacturing of cadmium salts, photographic emulsions, laboratory reagent and coloring glass. It is used for porcelain and as a flash powder in photography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Color White
Physical Form Crystals
Quantity 500 g
Assay Percent Range 98.5% min.
Solubility Information Highly soluble in water. Soluble in alcohol,acetone,and ammonia.
Formula Weight 308.47 (236.41 Anhydrous)
Odor Weak
Density 2.45 g/cm3
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX05G003 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number012228.22,012228.36
Lot NumberQ07I004 Certificate TypeCertificate of Analysis Date14/04/2022 Catalog Number012228.22,012228.36
Lot NumberZ07H018 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number012228.22,012228.36
Lot NumberC21Z038 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012228.22,012228.36
Lot NumberS04B020 Certificate TypeCertificate of Analysis Date17/11/2021 Catalog Number012228.22,012228.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium nitrate tetrahydrate, 98.5% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.