missing translation for 'onlineSavingsMsg'
Learn More
Cadmium nitrate tetrahydrate, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 11368888

Cadmium nitrate tetrahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 11368888
50 g
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy

Product Code. 11368888

Brand: Thermo Scientific Alfa Aesar 041732.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium nitrate tetrahydrate is used in Photographic emulsions, coloring glass and porcelain, laboratory reagent, and the production of other cadmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

Boiling Point 132°C
Melting Point 59°C
Physical Form Crystalline
Quantity 50 g
Assay Percent Range 99.9% (Metals basis)
UN Number UN3087
Merck Index 14,1622
Solubility Information Soluble in water.(109g/100ml)at 0 C and (326g/100ml)at 60 C.Soluble in ammonia,acetone and alcohol.
Formula Weight 308.47 (236.41 Anhydrous)
Percent Purity 99.9%
Odor Weak
Density 2.45 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium nitrate tetrahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ19K021 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number041732.30,041732.18
Lot NumberW23B038 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041732.30,041732.18
Lot NumberF03P08 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041732.30,041732.18
Lot NumberX12B004 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number041732.30,041732.18
Lot NumberF10M36 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number041732.30,041732.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium nitrate tetrahydrate, 99.9% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.