missing translation for 'onlineSavingsMsg'
Learn More

Cadmium nitrate tetrahydrate, 99.9% (metals basis), Thermo Scientific Chemicals

£80.20 - £257.28

Chemical Identifiers

CAS 10022-68-1
Molecular Formula CdH2N2O6
Molecular Weight (g/mol) 238.44
MDL Number MFCD00149626
InChI Key IZIPRBSCGMISKX-UHFFFAOYSA-N
Synonym cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
PubChem CID 56924536
ChEBI CHEBI:86156
IUPAC Name cadmium(2+);dinitrate;tetrahydrate
SMILES [Cd++].O[N+]([O-])=O.O[N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11368888
View Documents
Thermo Scientific Alfa Aesar
041732.18
50 g
£80.20
50g
Please to purchase this item. Need a web account? Register with us today!
11358888
View Documents
Thermo Scientific Alfa Aesar
041732.30
250 g
£257.28
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cadmium nitrate tetrahydrate is used in Photographic emulsions, coloring glass and porcelain, laboratory reagent, and the production of other cadmium compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10022-68-1
238.44
IZIPRBSCGMISKX-UHFFFAOYSA-N
56924536
cadmium(2+);dinitrate;tetrahydrate
CdH2N2O6
MFCD00149626
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
CHEBI:86156
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O

Specifications

132°C
10022-68-1
99.9% (Metals basis)
MFCD00149626
14,1622
Soluble in water.(109g/100ml)at 0 C and (326g/100ml)at 60 C.Soluble in ammonia,acetone and alcohol.
[Cd++].O[N+]([O-])=O.O[N+]([O-])=O
238.44
CHEBI:86156
99.9%
2.45 g/mL
Cadmium nitrate tetrahydrate
59°C
Crystalline
CdH2N2O6
UN3087
cadmium nitrate tetrahydrate, nitric acid, cadmium salt, tetrahydrate, cd no3 2.4h2o, acmc-20aljb, ksc174c1h, cadmium nitrate-water 1/4, cadmiumnitratetetrahydrate, cadmium ii nitrate tetrahydrate, nitric acid, cadmium salt tetrahydrate
IZIPRBSCGMISKX-UHFFFAOYSA-N
cadmium(2+);dinitrate;tetrahydrate
56924536
308.47 (236.41 Anhydrous)
Weak
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H350-H272-H312-H332
Toxic if swallowed.
May cause cancer.
May intensify fire
oxidizer.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P260-P264b-P270-P271-P281-P284-P301+P310-P302+P352-P304+P340-P308+P313-P310-P312-P330-P363-P501c

H301-H312-H330-H340-H350-H360-H372

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 233-710-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only