missing translation for 'onlineSavingsMsg'
Learn More
Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals Product Code.: 15289716

Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals

Product Code. 15289716
5 kg
Click to view available options
Quantity:
5 kg
This item is not returnable. View return policy

Product Code. 15289716

Brand: Thermo Scientific Acros 223790050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 1385.0°C
CAS 1306-19-0
Color Brown to Brown-Red
Physical Form Powder
Quantity 5 kg
Assay Percent Range 98.5% min. (Complexometry)
Molecular Formula CdO
Linear Formula CdO
MDL Number MFCD00010921
Merck Index 15,1628
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids and alkalies
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
SMILES [O--].[Cd++]
IUPAC Name cadmium(2+) oxidandiide
Molecular Weight (g/mol) 128.41
PubChem CID 14782
Formula Weight 128.41
Percent Purity 99%
Grade Pure
Chemical Name or Material Cadmium oxide, Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467624 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number223792500,223790050,223790010
Lot NumberA0465241 Certificate TypeCertificate of Analysis Date06/08/2024 Catalog Number223792500,223790050,223795000,223790010
Lot NumberA0462510 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number223792500,223790050,223795000,223790010
Lot NumberA0459802 Certificate TypeCertificate of Analysis Date01/02/2024 Catalog Number223792500,223790050,223795000,223790010
Lot NumberA0458488 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number223792500,223790050,223795000,223790010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium oxide, brown
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350-May cause cancer.
  • H341-Suspected of causing genetic defects.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium oxide, 99%, Pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.