missing translation for 'onlineSavingsMsg'
Learn More

Cadmium perchlorate hexahydrate, Thermo Scientific Chemicals

£58.19 - £235.17

Chemical Identifiers

CAS 10326-28-0
Molecular Formula CdCl2H12O14
Molecular Weight (g/mol) 419.396
MDL Number MFCD00015992
InChI Key PQJZOTJHMRJOEM-UHFFFAOYSA-L
Synonym cadmium perchlorate hexahydrate, acmc-1c6hb, cd.2clo4.6h2o, cadmium ii perchlorate hexahydrate
PubChem CID 71317310
IUPAC Name cadmium(2+);diperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Cd+2]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342028
SDS View Documents
Thermo Scientific Alfa Aesar
012936.18
50 g
£58.19
50g
Please to purchase this item. Need a web account? Register with us today!
11332028
SDS View Documents
Thermo Scientific Alfa Aesar
012936.30
250 g
£235.17
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is employed as intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data sheets.

P210-P220-P221-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P370+P378q-P501c

H272-H302+H312+H332

missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: II

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium perchlorate hexahydrate, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.