missing translation for 'onlineSavingsMsg'
Learn More
Cadmium powder, -200 mesh, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11331129

Cadmium powder, -200 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11331129
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11331129

Brand: Thermo Scientific Alfa Aesar 011353.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
SMILES [Cd]

Specifications

Color White
UN Number UN3179
Quantity 5 g
IUPAC Name cadmium(2+) ion
Percent Purity 99.99%
Mesh Size −200 Mesh
Form Solid
Assay (metals basis)
Chemical Name or Material Cadmium powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX16J053 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog Number011353.22,011353.14,011353.06
Lot NumberR18G005 Certificate TypeCertificate of Analysis Date25/05/2022 Catalog Number011348.14,011348.36,011348.22,011353.06
Lot NumberN18I027 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number011348.14,011348.36,011348.22,011353.06
Lot NumberJ18W012 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number011353.22,011355.03,011353.14,011353.06
Lot NumberR30D053 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number011353.22,011355.03,011353.14,011353.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium powder, -200 mesh, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.