missing translation for 'onlineSavingsMsg'
Learn More

Cadmium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11361988
100 g
Click to view available options
Quantity:
10 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Cadmium powder, 99.5%, Cd, CAS Number-7440-43-9, ccris 112, unii-00bh33gngh, compounds, cadmium, elemental, cadmium and compounds, 00bh33gngh, kadmium, kadmium german, colloidal, hsdb 282, 500g, (metals basis), CHEBI:22977, Silver-White

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Color Silver-White
UN Number UN3179
Quantity 100 g
Percent Purity 99.5%
Mesh Size −325 Mesh
Form Solid
Assay (metals basis)
Chemical Name or Material Cadmium powder

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.