missing translation for 'onlineSavingsMsg'
Learn More
Cadmium rod, 8mm (0.3in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11304788

Cadmium rod, 8mm (0.3in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11304788
100 mm
Click to view available options
Quantity:
100 mm
5 x 100 mm
This item is not returnable. View return policy

Product Code. 11304788

Brand: Thermo Scientific Alfa Aesar 043116.DM

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow MoreShow Less
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Quantity 100 mm
Percent Purity 99.999%
Grade Puratronic™
Weight ≈0.43g/mm
Form Rod
Assay (metals basis)
Chemical Name or Material Cadmium rod, 8mm (0.3 in.) dia.

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN14K008 Certificate TypeCertificate of Analysis Date05/03/2024 Catalog Number043116.DM,043116.HR
Lot NumberS01I028 Certificate TypeCertificate of Analysis Date22/08/2022 Catalog Number043116.DM,043116.HR
Lot NumberX22H005 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043116.DM,043116.HR
Lot NumberE19N51 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043116.DM,043116.HR
Lot NumberC30L50 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number043116.DM,043116.HR

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium rod, 8mm (0.3in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.