missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Cadmium shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9997% (metals basis)
Klicka för att se tillgängliga alternativ
Quantity:
50 g
250 g
1 kg
Förpackningsstorlek:
1kg
250g
50g
Beskrivning
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifikationer
Specifikationer
| Color | Silver-White |
| Appearance | Silvery-white shot |
| Density | 8.65g/cm3 |
| Quantity | 50 g |
| Vapor Pressure | 0.00013 hPa |
| Assay Percent Range | 99.9997% (Metals basis) |
| Form | Shot |
| IUPAC Name | cadmium |
| Chemical Name or Material | Cadmium |
Säkerhet och hantering
Produktbeteckning
- Cadmium shot
- Danger
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cadmium
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering