missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£79.10 - £145.00
Chemical Identifiers
CAS | 7440-43-9, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Cd |
Molecular Weight (g/mol) | 112.41 |
MDL Number | MFCD00010914 |
InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
PubChem CID | 23973 |
ChEBI | CHEBI:22977 |
IUPAC Name | cadmium |
SMILES | [Cd] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
11909482
|
SPEX Certiprep
PLCD2-2Y |
125 mL | Triple-leached LDPE bottle |
£79.10
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10790375
|
SPEX Certiprep
PLCD2-2T |
250 mL | Triple-leached LDPE bottle |
£93.50
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10522333
|
SPEX Certiprep
PLCD2-2X |
500 mL | Triple-leached LDPE bottle |
£145.00
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cd | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-43-9, 7697-37-2, 7732-18-5 | |
112.41 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
CHEBI:22977 | |
[Cd] |
Cd | |
MFCD00010914 | |
23973 | |
cadmium |
Specifications
7440-43-9, 7697-37-2, 7732-18-5 | |
0.1, 2.0, 97.9 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
Cd | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
cadmium | |
23973 | |
Assurance | |
2% HNO3 |
0.1, 2.0, 97.9 | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00010914 | |
[Cd] | |
112.41 | |
CHEBI:22977 | |
Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mLPLCD2-2Y, -2T, -2X 10,000 PLCD2-3Y, -3X 2-5% HCl 1000μg/mL PLCD1-2X |
|
Cadmium Standard |
Safety and Handling
Causes skin irritation, serious eye irritation. May cause cancer. Suspected of damaging fertility or the unborn child.
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only