missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium sulfate, ACS reagent, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 1000.0°C |
Quantity | 100 g |
Linear Formula | CdSO4 |
Merck Index | 15,1632 |
Solubility Information | Solubility in water: 1130g/L (20°C). Other solubilities: insoluble in alcohol, acids, acetone and ammonia |
Formula Weight | 208.46 |
Grade | ACS Reagent |
Packaging | Glass bottle |
Chemical Name or Material | Cadmium sulfate |
Safety and Handling
Product Identifier
- Cadmium sulphate
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cadmium sulphate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'