missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium Sulfate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
100 g
500 g
Specifications
Specifications
Melting Point | 41°C |
Color | White |
pH | 4.5 to 6.0 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 769.51g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Cadmium sulfate, 8/3-hydrate
- Danger
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Acute toxicity Category 2
- Acute toxicity Category 3
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H350-May cause cancer.
- H340-May cause genetic defects.
- H360FD-May damage fertility. May damage the unborn child.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P310-Immediately call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'