missing translation for 'onlineSavingsMsg'
Learn More

Cadmium sulfate octahydrate, ACS, 98.0-102.0%

Product Code. p-4407304
Click to view available options
Quantity:
25 g
250 g
1 kg
Unit Size:
1kg
250g
25g
This item is not returnable. View return policy

Product Code. 11381709

Brand: Thermo Scientific Alfa Aesar 012340.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used in Standard Weston Cell, as electrolytes in electroplating, in phosphors, in manufacture of standard cadmium elements, and as a catalyst in Marsh test for AS | CAS: 7790-84-3 | 3CdSO4·8H2O

Cadmium sulfate mainly used in electroplating of cadmium, which is used in electronic circuits. It is used as a precursor for cadmium based pigments such as cadmium sulfide and pharmaceutical intermediates. It is used in the standard weston cell.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-84-3
MDL Number MFCD00151655

Specifications

Melting Point >40°C to H2O
Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98.0 to 102.0%
Molecular Formula 3CdSO4·8H2O
Solubility Information Highly soluble in water. Partially soluble in methanol and ethyl acetate. Insoluble in ethanol.
Formula Weight 769.50 (208.46 Anhydrous)
Grade ACS
Refractive Index 1.565
Appearance White crystalline
Density 3.08 g/mL
Chemical Name or Material Cadmium sulfate octahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium sulfate octahydrate, ACS
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 1B
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H330-Fatal if inhaled.
  • H340-May cause genetic defects.
  • H350-May cause cancer.
  • H360FD-May damage fertility. May damage the unborn child.
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cadmium sulfate octahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.