Learn More
Cadmium sulfide, 98%, Thermo Scientific Chemicals
£28.44 - £403.01
Chemical Identifiers
CAS | 1306-23-6 |
---|---|
Molecular Formula | CdS |
Molecular Weight (g/mol) | 144.47 |
MDL Number | MFCD00010922 |
InChI Key | FRLJSGOEGLARCA-UHFFFAOYSA-N |
Synonym | cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellowShow More |
PubChem CID | 14783 |
ChEBI | CHEBI:50833 |
SMILES | [S--].[Cd++] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11461918
|
Thermo Scientific Alfa Aesar
A14544.22 |
100 g |
£28.44
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11471918
|
Thermo Scientific Alfa Aesar
A14544.36 |
500 g |
£102.51
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11481918
|
Thermo Scientific Alfa Aesar
A14544.0E |
2500 g |
£403.01
2500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCadmium sulfide is used as a pigment in the chemical industry; for manufacturing of photoresistors, optical-electronic pairs, photodiodes, solar batteries and phosphors. Single crystals of cadmium sulfide are used for the preparation of optical devices operating in the infrared regaion of spectrum. Cadmium sulfide is a direct band gap semiconductor with a band gap band from 2.42 to 2.57 eV.
Solubility
Insoluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and acids.
Chemical Identifiers
1306-23-6 | |
144.47 | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
14783 | |
[S--].[Cd++] |
CdS | |
MFCD00010922 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellowShow More | |
CHEBI:50833 |
Specifications
980°C (sublimation) | |
CdS | |
UN3077 | |
cadmium sulfide, cadmium sulphide, capsebon, orange cadmium, cadmopur yellow, jaune brilliant, aurora yellow, cadmium orange, cadmium yellow, ferro yellow | |
FRLJSGOEGLARCA-UHFFFAOYSA-N | |
sulfanylidenecadmium | |
14783 | |
144.46 | |
Odorless | |
Cadmium sulfide |
1306-23-6 | |
MFCD00010922 | |
14,1628 | |
Insoluble in water. | |
[S--].[Cd++] | |
144.47 | |
CHEBI:50833 | |
98% | |
4.82 |
Safety and Handling
GHS H Statement
H350-H372-H341-H361-H302
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
P201-P202-P260-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
H302-H341-H350-H361fd-H372
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III
missing translation for 'einecsNumber' : 215-147-8
missing translation for 'rtecsNumber' : EV3150000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cadmium sulphide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 4
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 2
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Cadmium sulphide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.