missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
Description
The main application of cadmium sulfide is as a pigment. Cadmium sulfide is also used in the production of solar cells. Used as light dependent resistors/photo resistors for light sensors. Used in MOCVD, sputtering and electrochemical deposition. In screen printing, it is used as a slurry containing dispersed cadmium sulfide. It is also used in dandruff shampoos.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 980°C (sublimation) |
Color | Orange |
Physical Form | Powder/Granules |
Quantity | 50 g |
UN Number | UN3077 |
Merck Index | 14,1628 |
Solubility Information | Soluble in acid,very slightly soluble in ammonium hydroxide. |
Formula Weight | 144.46 |
Percent Purity | 99.999% |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'