missing translation for 'onlineSavingsMsg'
Learn More

Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. p-7076482
Click to view available options
Quantity:
1 g
25 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1g
25g
This item is not returnable. View return policy

Product Code. 15402277

Brand: Thermo Scientific Alfa Aesar 045457.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Used to make thin film solar cells.

Cadmium telluride is used to make thin film solar cells. It can be alloyed with mercury to make a versatile infrared detector material (HgCdTe). It is used as an infrared optical material for optical windows and lenses and is proven to provide a good performance across a wide range of temperatures. It is also applied for electro-optic modulators and has the greatest electro-optic coefficient of the linear electro-optic effect among II-VI compound crystals. It can be doped with chlorine, which is used as a radiation detector for x-rays, gamma rays, beta particles and alpha particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-25-8
Molecular Formula CdTe
Molecular Weight (g/mol) 240.01
MDL Number MFCD00015998
InChI Key RPPBZEBXAAZZJH-UHFFFAOYSA-N
Synonym cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basis
PubChem CID 91501
IUPAC Name tellanylidenecadmium
SMILES [Cd]=[Te]

Specifications

Melting Point 1041°C
Physical Form Polycrystalline Pieces
Quantity 1 g
UN Number UN3077
Solubility Information Insoluble in water.
Formula Weight 240
Percent Purity 99.999%
Packaging Plastic bottle
Specific Gravity 6.2
Density 6.2
Chemical Name or Material Cadmium telluride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.