missing translation for 'onlineSavingsMsg'
Learn More
Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals Product Code.: 11367917

Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals

Product Code. 11367917
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11367917

Brand: Thermo Scientific Alfa Aesar 014365.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles. .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-25-8
Molecular Formula CdTe
Molecular Weight (g/mol) 240.01
MDL Number MFCD00015998
InChI Key RPPBZEBXAAZZJH-UHFFFAOYSA-N
Synonym cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basisShow MoreShow Less
PubChem CID 91501
IUPAC Name tellanylidenecadmium
SMILES [Cd]=[Te]

Specifications

Melting Point 1041°C
Color Black
Physical Form Polycrystalline Lump
Quantity 100 g
Assay Percent Range 99.999% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 240
Odor Odorless
Density 6.2
Chemical Name or Material Cadmium telluride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD29Z080 Certificate TypeCertificate of Analysis Date20/01/2025 Catalog Number014365.22,014365.14
Lot NumberS17K021 Certificate TypeCertificate of Analysis Date04/07/2024 Catalog Number014365.22,014365.14
Lot NumberT24J025 Certificate TypeCertificate of Analysis Date14/08/2023 Catalog Number014365.22,014365.14
Lot NumberP06B010 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number014365.22,014365.14,014367.03,014365.06
Lot NumberX11H065 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number014365.22,014365.14,014367.03,014365.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium telluride, Black polycrystalline lump, 99.999% (Metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.