missing translation for 'onlineSavingsMsg'
Learn More

Cadmium wire, 0.3mm (0.013 in.) dia., Puratronic™, 99.999% (metals basis), Thermo Scientific™

Product Code. 11308688
Click to view available options
Quantity:
10 m
2 m

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Quantity 2 m
Percent Purity 99.999%
Grade Puratronic™
Weight ≈0.61g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Cadmium wire, 0.3mm (0.013 in.) dia.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'