missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caesium Chloride, 99.5+%, Certified AR for Analysis, Fisher Chemical™
ClCs, CAS Number-7647-17-8, 1kg, 1290 deg.C, White, 168.36g/mol, 646 deg.C, 10955, Plastic powder jar, 5 to 9, Powder Solid
£101.00 - £1975.00
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
IUPAC Name | caesium(1+) chloride |
SMILES | [Cl-].[Cs+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10775321
|
Fisher Chemical
C/0680/46 |
25 g |
£101.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10213710
|
Fisher Chemical
C/0680/48 |
100 g |
£323.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10785321
|
Fisher Chemical
C/0680/50 |
250 g |
£660.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10588030
|
Fisher Chemical
C/0680/60 |
1 kg |
£1975.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ClCs, CAS Number-7647-17-8, 1kg, 1290 deg.C, White, 168.36g/mol, 646 deg.C, 10955, Plastic powder jar, 5 to 9, Powder Solid
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
[Cl-].[Cs+] |
ClCs | |
MFCD00010955 | |
caesium(1+) chloride |
Specifications
1290°C | |
7647-17-8 | |
5 to 9 | |
ClCs | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
caesium(1+) chloride | |
168.36g/mol |
646°C | |
White | |
Powder/Solid | |
MFCD00010955 | |
[Cl-].[Cs+] | |
168.36 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Cesium chloride
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.