missing translation for 'onlineSavingsMsg'
Learn More
Caffeine, 99%, Thermo Scientific Chemicals Product Code.: 11364569

Caffeine, 99%, Thermo Scientific Chemicals

Product Code. 11364569
100 g
Click to view available options
Quantity:
100 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11364569

Brand: Thermo Scientific Alfa Aesar A10431.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This material is traded for research purposes ONLY. Thermo Fisher does NOT certify this material as suitable for pharmaceutical, cosmetic, veterinary or food use. Caffeine has inhibitory action against lung and colon tumorigenses and virus induction.Caffeine is used to treat and prevent bronchopulmonary dysplasia in premature infants. It is also used for the treatment of apnea of prematurity and orthostatic hypotension. It is widely used as an additive in food, cosmetics, pharmaceuticals and beverages. It confers a protective effect against some diseases including Parkinson's disease and certain types of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in pyrrole, tetrahydrofuran+ water mixture and ethyl acetate. Slightly soluble in water and ethanol.

Notes
Store in cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-08-2
Molecular Formula C8H10N4O2
Molecular Weight (g/mol) 194.19
MDL Number MFCD00005758
InChI Key RYYVLZVUVIJVGH-UHFFFAOYSA-N
Synonym caffeine, 1,3,7-trimethylxanthine, guaranine, thein, cafeina, methyltheobromine, koffein, mateina, theine, alert-pep
PubChem CID 2519
ChEBI CHEBI:27732
SMILES CN1C=NC2=C1C(=O)N(C)C(=O)N2C

Specifications

Melting Point 234°C to 236°C
Density 1.23
Assay Percent Range 99%
Quantity 100 g
UN Number UN1544
Beilstein 17705
Merck Index 14,1636
Solubility Information Soluble in pyrrole,tetrahydrofuran+water mixture and ethyl acetate; Slightly soluble in water and ethanol.
IUPAC Name 1,3,7-trimethylpurine-2,6-dione
Formula Weight 194.19
Percent Purity 99%
Chemical Name or Material Caffeine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberR06K031 Certificate TypeCertificate of Analysis Date15/05/2024 Catalog NumberA10431.0B,A10431.22,A10431.30
Lot NumberP18K026 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberA10431.0B,A10431.22,A10431.30
Lot NumberP04K034 Certificate TypeCertificate of Analysis Date11/04/2024 Catalog NumberA10431.0B,A10431.22,A10431.30
Lot NumberP24J032 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberA10431.22
Lot Number10245599 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog NumberA10431.0B,A10431.22,A10431.30
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Caffeine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.