missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caffeine, Extra Pure, SLR, Fisher Chemical
Click to view available options
100g, Plastic powder jar
Specifications
Specifications
CAS | 58-08-2 |
Melting Point | 234°C |
Color | White |
pH | 5.5 to 6.5 |
Packaging | Plastic powder jar |
Molecular Formula | C8H10N4O2 |
MDL Number | 5758 |
Quantity | 100 g |
Formula Weight | 194.19g/mol |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Caffeine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction