missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcein Disodium Salt, Honeywell Fluka™
Click to view available options
Quantity:
1 g
5 g
100 g
Packaging:
Glass Bottle
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
UN Number | NONH for all modes of transport |
Formula Weight | 666.5g/mol |
Packaging | Glass Bottle |
Color | Red |
Quantity | 5 g |
Physical Form | Powder |
Chemical Name or Material | Calcein Disodium Salt, For complexometry |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'