Learn More
Calcifediol-d6, MedChemExpress
MedChemExpress Calcifediol-d6 is the deuterium labeled Calcifediol. Calcifediol, a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.
£1201.53 - £4806.12
Specifications
Chemical Name or Material | Calcifediol-d6 |
---|---|
CAS | 78782-98-6 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C27H38D6O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16471339
|
Medchem Express
HY-13332 |
1 mg |
£1201.53
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16468347
|
Medchem Express
HY-13332 |
5 mg |
£4806.12
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcifediol-d6 is the deuterium labeled Calcifediol. Calcifediol, a major circulating metabolite of vitamin D3, is a potent VDR inhibitor.Specifications
Calcifediol-d6 | |
White | |
C27H38D6O2 | |
406.67 | |
98.0% | |
Metabolism-sugar/lipid metabolism |
78782-98-6 | |
Research | |
O[C@@H](CCC/1=C)CC1=C\C=C2[C@@](CC[C@@H]3[C@@H](CCCC(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)([H])[C@]3(C)CCC\2 | |
406.67 | |
Research | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Recommended Storage : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.