missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcitriol, Tocris Bioscience™
Click to view available options
Quantity:
50 μg
Description
Active metabolite of vitamin D3 that activates the vitamin D receptor (VDR). Displays calcemic actions; stimulates intestinal and renal Ca2+ absorption and regulates bone Ca2+ turnover. Exhibits antitumor activity; inhibits in vivo and in vitro cell proliferation in a wide range of cells including breast, prostate, colon, skin and brain carcinomas and myeloid leukemia cells.
Specifications
Specifications
CAS | 32222-06-3 |
Molecular Formula | C27H44O3 |
MDL Number | MFCD00867079,MFCD11656129 |
Quantity | 50 μg |
Synonym | calcitriol, rocaltrol, calcijex, topitriol, 1alpha,25-dihydroxyvitamin d3, silkis, soltriol, 1alpha,25-dihydroxycholecalciferol, calcitriolum, dihydroxyvitamin d3 |
InChI Key | GMRQFYUYWCNGIN-NKMMMXOESA-N |
SMILES | C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C |
IUPAC Name | (1R,3S,5Z)-5-{2-[(1R,3aS,4E,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol |
Molecular Weight (g/mol) | 416.65 |
PubChem CID | 5280453 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction