missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium 1000 ppm Ion Selective Electrode Standard, Reagecon
Can be used at suitable concentrations to calibrate or control measurements that use ion selective electrodes (ISE) as the sensing technology.
Brand: Reagecon ISECA5
Code : A0
Additional Details : Weight : 0.45000kg
Description
- Ion Selective Electrode Reference Standard, Ion Selective Electrode Reference Material
- Certified
- NIST traceable
- Accurate to ± 0.5%
- Safe, compatible with all types of ISE Electrodes
- Can be used as a calibrant for other techniques
- Can be used for method validation or instrument qualification
- Aqueous matrix
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Accuracy ± 0.5% | |
HDPE Bottle GL45 | |
Calcium |
1000 ppm ± 0.5% | |
500 mL | |
Liquid |