missing translation for 'onlineSavingsMsg'
Learn More
Calcium, 99%, granular, Thermo Scientific Chemicals Product Code.: 10579253

Calcium, 99%, granular, Thermo Scientific Chemicals

Product Code. 10579253
5 g, Plastic Bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10579253

Brand: Thermo Scientific Acros 201180050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]

Specifications

Color Gray
Melting Point 845.0°C
Boiling Point 1484.0°C
Assay Percent Range 99%
Linear Formula Ca
Quantity 5 g
Fieser 05,89; 09,1634; 14,71
Merck Index 15, 1648
Formula Weight 40.07
Percent Purity 99%
Packaging Plastic Bottle
Form Granular
Chemical Name or Material Calcium, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2198973 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number201180050,201185000,201181000,201180025
Lot Number2362626 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number201180050,201185000,201181000,201180025
Lot Number2050993 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number201180050,201185000,201181000,201180025
Lot Number2048643 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number201180050,201185000,201181000,201180025
Lot Number2228294 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog Number201180050,201185000,201181000,201180025
compliance-icons
Product Identifier
  • Calcium
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P223-Do not allow contact with water.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P370+P378-In case of fire: Use carbon dioxide to extinguish.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium, 99%, granular, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.