missing translation for 'onlineSavingsMsg'
Learn More

Calcium acetate monohydrate, ACS, 99.0% min

Product Code. p-4407308
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 11316798

Brand: Thermo Scientific Alfa Aesar 013446.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5743-26-0 | C4H8CaO5 | 176.181 g/mol

Calcium acetate monohydrate is used to precipitate oxalates in solution. It is used to control the phosphate levels in blood, thereby controlling the bone problems. It is also involved in the neutralization of fluoride in water. Further, it is used as a food additive mainly in candy products. In addition to this, it serves as a stabilizer, buffer and sequestrant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H8CaO5
Molecular Weight (g/mol) 176.181
MDL Number MFCD00150019
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Melting Point (decomposition)
Color White
Physical Form Powder
Quantity 250 g
Assay Percent Range 99%
Beilstein 3692527
Solubility Information Soluble in water. Slightly soluble in methanol and hydrazine. Insoluble in ethanol,acetone and benzene.
Formula Weight 176.18 (158.17 Anhydrous)
Grade ACS
Sensitivity Hygroscopic
Density 1.5 g/cm3
Chemical Name or Material Calcium acetate monohydrate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.