missing translation for 'onlineSavingsMsg'
Learn More

Calcium alpha-D-isosaccharinate, 98% (Assay), Thermo Scientific Chemicals

£25.00 - £1393.00

Chemical Identifiers

CAS 84324-95-8
Molecular Formula C12H22CaO12
MDL Number MFCD04039767
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11410710
View Documents
Thermo Scientific Alfa Aesar
044371.01
0.25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11468540
View Documents
Thermo Scientific Alfa Aesar
044371.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11458540
View Documents
Thermo Scientific Alfa Aesar
044371.06
5 g
£287.00
5g
Please to purchase this item. Need a web account? Register with us today!
11478540
View Documents
Thermo Scientific Alfa Aesar
044371.14
25 g
£1393.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

m alpha-D-isosaccharinate is used to prevent or treat low blood calcium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

84324-95-8
C12H22CaO12
MFCD04039767

Specifications

84324-95-8
Crystalline
[CO2C(OH)(CH2OH)CH2HCOHCH2OH]2Ca
398.38
(Assay)
Brown
C12H22CaO12
MFCD04039767
98%
Calcium alpha-D-isosaccharinate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only