missing translation for 'onlineSavingsMsg'
Learn More

Calcium aluminum oxide, 99% (metals basis)

Product Code. p-4781833
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11328077

Brand: Thermo Scientific Alfa Aesar 022646.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12042-68-1 | Al2CaO4 | 158.037 g/mol

Calcium aluminum oxide is used as a precursor in the production of naphtha steam reforming catalysts, refinery gas steam reforming catalysts, aluminum chlorohydrate, secondary hydrocarbon steam reforming catalysts. It is also used in the production of refractory and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12042-68-1
Molecular Formula Al2CaO4
Molecular Weight (g/mol) 158.037
MDL Number MFCD00049722
InChI Key XFWJKVMFIVXPKK-UHFFFAOYSA-N
Synonym calcium aluminate, calcium aluminum oxide, calcium dialumanoylolate, dialuminium calcium tetraoxide, calcium dialuminate, aluminum calcium oxide, 2alo2.ca, calcium oxido oxo alumane, calcium 2+ dialumanoylolate, calcium aluminate 5g
PubChem CID 159415
IUPAC Name calcium;oxido(oxo)alumane
SMILES [O-][Al]=O.[O-][Al]=O.[Ca+2]

Specifications

Melting Point 1600°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99% (Metals basis)
Solubility Information Insoluble in water.
Formula Weight 158.02
Odor Odorless
Sensitivity Hygroscopic
Density 2.981
Chemical Name or Material Calcium aluminum oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.