missing translation for 'onlineSavingsMsg'
Learn More

Calcium boride, 95+%

Product Code. 11368868
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11368868

Brand: Thermo Scientific Alfa Aesar 041721.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12007-99-7 | B6Ca | 104.938 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12007-99-7
Molecular Formula B6Ca
Molecular Weight (g/mol) 104.938
MDL Number MFCD00148904
InChI Key AWVIDOFQNLOVRC-UHFFFAOYSA-N
Synonym calcium boride, calcium hexaboride, b6.ca, calcium hexaboride,-200 mesh, calcium pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide, calcium 2+ pentacyclo 3.1.0.0^ 1,3 .0^ 2,4 .0^ 4,6 hexaborane-1,4-diuide
PubChem CID 16212529
SMILES B12B3[B-]14B5[B-]23B45.[Ca+2]

Specifications

Melting Point 2235°C
Physical Form Powder
Quantity 1 kg
Solubility Information Insoluble in water. Dissolves slowly in acids
Formula Weight 104.95
Percent Purity ≥95%
Density 2.30 g/mL
Chemical Name or Material Calcium boride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.