missing translation for 'onlineSavingsMsg'
Learn More

Calcium bromide hydrate, 95%

Product Code. 11333487
Click to view available options
Quantity:
5 g
100 g
500 g
Unit Size:
100g
500g
5g
This item is not returnable. View return policy

Product Code. 11333487

Brand: Thermo Scientific Alfa Aesar 018820.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 71626-99-8 | Br2Ca | 199.89 g/mol

Calcium bromide hydrate is used as freezing mixtures, food preservatives and photography. Its aqueous solution is used in drilling fluids and fire retardants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71626-99-8
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.89
MDL Number MFCD00149608
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
IUPAC Name calcium dibromide
SMILES [Ca++].[Br-].[Br-]

Specifications

Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 95.0%
Solubility Information Soluble in water.
Formula Weight 199.90 (Anhydrous)
Odor Odorless
Appearance White powder
Chemical Name or Material Calcium bromide hydrate
compliance-icons
Product Identifier
  • Calcium bromide hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Calcium bromide, hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.