missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium bromide hydrate, for analysis
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
500g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 730.0°C |
| Color | White |
| Physical Form | Crystalline Powder and/or Chunks |
| Quantity | 2.5 kg |
| Linear Formula | CaBr2·xH2O |
| Solubility Information | Solubility in water: 1420g/L (20°C). Other solubilities: soluble in methanol,ethanol |
| Formula Weight | 199.9 |
| Percent Purity | ≥83.0% (Complexometry) (CaBr2) |
| Grade | Analytical |
| Packaging | Plastic bottle |
| Show More |
Safety and Handling
Product Identifier
- Calcium bromide hydrate
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : Calcium bromide, hydrate
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction