Learn More
Calcium carbide, 72-82%, Thermo Scientific Chemicals
£23.40 - £58.30
Chemical Identifiers
CAS | 75-20-7 |
---|---|
Molecular Formula | C2Ca |
Molecular Weight (g/mol) | 64.1 |
MDL Number | MFCD00010905 |
Synonym | calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10646124
|
Thermo Scientific Acros
389792500 |
250 g | Glass bottle |
£23.40
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10762284
|
Thermo Scientific Acros
389790010 |
1 kg | Glass bottle |
£58.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-20-7 | |
64.1 | |
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate |
C2Ca | |
MFCD00010905 |
Specifications
2300.0°C | |
75-20-7 | |
Lump | |
C2Ca | |
MFCD00010905 | |
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate | |
64.1 | |
72 to 82% | |
Calcium carbide |
2160.0°C | |
Gray | |
12 to 23% (CaO) (Complexometry) | |
CaC2 | |
15, 1658 | |
Solubility in water: hydrolyses | |
64.1 | |
Positive |
Safety and Handling
GHS H Statement
In contact with water releases flammable gases which may ignite spontaneously.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye damage.
GHS P Statement
Keep away from any possible contact with water,because of violent reaction and possible flash fire.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protecti
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-848-3
RUO – Research Use Only