Learn More
Calcium carbide, Thermo Scientific Chemicals
£52.86 - £182.91
Chemical Identifiers
CAS | 75-20-7 |
---|---|
Molecular Formula | C2Ca |
MDL Number | MFCD00010905 |
Synonym | calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337877
|
Thermo Scientific Alfa Aesar
014308.A1 |
1 kg |
£52.86
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327877
|
Thermo Scientific Alfa Aesar
014308.D9 |
5 x 1 kg |
£182.91
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium carbide is used as a desulfurizer, dehydrant of steel, fuel in steel making, powerful deoxidizer and as a source of acetylene gas. It is used as a starting material for the preparation of calcium cyanamide, ethylene, chloroprene rubber, acetic acid, dicyandiamide and cyanide acetate. It is used in carbide lamps, toy cannons such as the big-bang cannon and bamboo cannon. It is associated with calcium phosphide and used in floating, self-igniting naval signal flares. Further, it is involved in the reduction of copper sulfide to metallic copper.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
75-20-7 | |
C2Ca | |
MFCD00010905 | |
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate |
Specifications
2300°C | |
Gray to Black | |
C2Ca | |
3909011 | |
64.09 | |
Grayish black lump | |
Calcium carbide |
75-20-7 | |
Lump | |
MFCD00010905 | |
calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate | |
Moisture sensitive | |
2.22 g/cm3 |
Safety and Handling
P223-P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P335+P334-P362-P370-P378aa-P378ac-P501c
H260-H315-H318-H335
missing translation for 'dotInformation' : DOT Class: 4.3; Packaging Group: I
missing translation for 'einecsNumber' : 200-848-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Calcium carbide
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
Precautionary Statement
- P223-Do not allow contact with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P334-Immerse in cool water/wrap in wet bandages.
Supplemental information
- MIXTURE LIST-Contains : Calcium carbide, Calcium oxide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.