Learn More
Calcium carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Description
Calcium carbonate is used in the purification of iron, refining of sugar from sugar beet and flue gas desulfurization applications in fossil fuel power stations. It is used in blackboard chalk, printing and writing paper, paints, plastics, PVC cables, polypropylene compounds, thermosetting resins, adhesives, sealants and decorating fillers. It is an abrasive and an ingredient in many glazes. It is also used in the pharmaceutical industry as an inert filler for tablets and other pharmaceuticals. It is used as a base in many organic reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 800°C (decomposition) |
Color | White |
Physical Form | Powder |
Quantity | 500 g |
Assay Percent Range | 99.5% (Metal basis) |
Solubility Information | Very low solubility in water. |
Formula Weight | 100.09 |
Refractive Index | 1.6583 |
Appearance | White powder |
Density | 2.930 g/cm3 |
Show More |
RUO – Research Use Only