missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate, 99%, extra pure, Thermo Scientific Chemicals
£31.76 - £305.00
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15218877
|
Thermo Scientific Acros
450680010 |
1 kg |
£31.76
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15204258
|
Thermo Scientific Acros
450680050 |
5 kg |
£64.12
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15214258
|
Thermo Scientific Acros
450680250 |
25 kg |
£305.00
25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
825.0°C | |
98.5 to 101.5% (Complexometry) | |
CaCO3 | |
15,1659 | |
(in dilute HCl) Clear colorless to light yellow | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
99% | |
2% max. (200°C) |
471-34-1 | |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
100.09 | |
Extra Pure | |
Calcium carbonate |
Safety and Handling
missing translation for 'einecsNumber' : 207-439-9
Product Identifier
- Calcium carbonate
RUO – Research Use Only