missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, 99+%, ACS reagent

Product Code. 10463655 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10463655

Brand: Thermo Scientific Acros 423510025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 99% min. (on dried substance) (Complexometry)
Linear Formula CaCO3
Fieser 01,103; 02,57; 04,67; 05,89
Merck Index 15, 1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
IUPAC Name calcium;carbonate
Formula Weight 100.09
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Calcium carbonate, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.