missing translation for 'onlineSavingsMsg'
Learn More
Calcium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals Product Code.: 11347608

Calcium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals

Product Code. 11347608
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11347608

Brand: Thermo Scientific Alfa Aesar 033295.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium carbonate is used medicinally as a calcium supplement or as an antacid. It is also used for industrial purposes. Calcium carbonate is the most widely used mineral in the paper, plastics, paints and coatings industries. Hierarchical hollow calcium carbonate particles show a useful application as an anticancer drug carrier. It plays an essential role to catalyze alcoholysis of fats and oils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 99.0% min.
Solubility Information Soluble in acids. Insoluble in water.
IUPAC Name calcium carbonate
Formula Weight 100.09
Grade ACS
Refractive Index 1.6583
Appearance White powder
Density 2.93 g/cm3 at 20°C
Chemical Name or Material Calcium carbonate, For ACS analysis
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ19J008 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number033295.22,033295.36
Lot NumberM11J017 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number033295.22,033295.36
Lot NumberZ20I020 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number033295.22,033295.36
Lot NumberZ20I021 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number033295.22,033295.36
Lot NumberL11I06 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number033295.22,033295.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium carbonate, ACS, 99.0% min, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.