Learn More
Calcium carbonate, chelometric standard, ACS, 99.95-100.05%, Thermo Scientific Chemicals
Calcium carbonate, CCaO3, CAS Number-471-34-1, aeromatt, akadama, aragonite, calcite, calcium carbonate, calofort u, carbonic acid calcium salt 1:1, chalk, limestone, marble, 500g, White powder, 99.95-100.05%, CHEBI:3311, 2.93, 207-439-9, 100.09, ACS, VTYYLEPIZMXCLO-UHFFFAOYSA-L, calcium;carbonate
£41.10 - £115.00
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11387858
|
Thermo Scientific Alfa Aesar
036399.22 |
100 g |
£41.10
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397858
|
Thermo Scientific Alfa Aesar
036399.36 |
500 g |
£115.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium carbonate is a dietary supplement used when the quantity of calcium taken in the diet is not enough. Used as an antacid to relieve heartburn, acid indigestion and upset belly. Practiced in the expression industry. Also widely used as a filler in plastics and is used in metallurgy of iron.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
800°C (decomposition) | |
White | |
99.95-100.05% | |
MFCD00010906 | |
Insoluble in water but soluble in acids. | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
ACS | |
White powder | |
Calcium carbonate, For analysis ACS |
471-34-1 | |
Powder | |
CCaO3 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium carbonate | |
10112 | |
100.09 | |
1.6583 | |
2.93 g/cm3 at 20°C |
Safety and Handling
missing translation for 'einecsNumber' : 207-439-9
missing translation for 'rtecsNumber' : FF9335000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only