missing translation for 'onlineSavingsMsg'
Learn More

Calcium carbonate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

£49.04 - £444.21

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15487485
View Documents
Thermo Scientific Alfa Aesar
047235.06
5 g
£49.04
5g
Please to purchase this item. Need a web account? Register with us today!
15497485
View Documents
Thermo Scientific Alfa Aesar
047235.14
25 g
£150.75
25g
Please to purchase this item. Need a web account? Register with us today!
15407495
View Documents
Thermo Scientific Alfa Aesar
047235.22
100 g
£444.21
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applied as neutralizing agent, filler, and extender in rubber, plastics, paints, opacifying agent in paper, putty, antacid, Portland cement, sulfur dioxide, removal from stack gases, allurgical flux, analytical chemistry, carbon dioxide generation, and determining halogens in organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

471-34-1
100.09
VTYYLEPIZMXCLO-UHFFFAOYSA-L
10112
[Ca++].[O-]C([O-])=O
CCaO3
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
CHEBI:3311

Specifications

800°C (decomposition)
White
CCaO3
14,1657
Soluble in dilute acids (5 M HCl: 0.1 M at 20°C,clear,colorless). Insoluble in water.
[Ca++].[O-]C([O-])=O
100.09
CHEBI:3311
99.999%
Odorless
(metals basis)
471-34-1
Powder
MFCD00010906
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
VTYYLEPIZMXCLO-UHFFFAOYSA-L
calcium;carbonate
10112
100.09
Puratronic™
2.930 g/mL
Calcium carbonate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 207-439-9

missing translation for 'rtecsNumber' : FF9335000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only