missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium chloride, 96%, for analysis, granules, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | >1600.0°C |
Melting Point | 782.0°C |
Physical Form | Granules |
Quantity | 500 g |
Assay Percent Range | 95% min. (Complexometry) |
Linear Formula | CaCl2 |
Fieser | 01,104; 04,368 |
Merck Index | 15, 1661 |
Solubility Information | Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol |
Formula Weight | 110.99 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Calcium chloride
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'