missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, anhydrous, 93%, Thermo Scientific Chemicals

£43.42 - £119.60

Chemische Identifikatoren

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
Produktcode Marke Quantity Preis Menge & Verfügbarkeit  
11301689
Unterlagen ansehen
Thermo Scientific Alfa Aesar
012316.A1
1 kg
£43.42
1kg
Please to purchase this item. Need a web account? Register with us today!
11311689
Unterlagen ansehen
Thermo Scientific Alfa Aesar
012316.A7
5 kg
£119.60
5kg
Please to purchase this item. Need a web account? Register with us today!
Beschreibung

Beschreibung

It is used as a drying and dehydrating agent for organic liquids and gases, and for solids in desiccators. Common applications include brine for refrigeration plants, ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranate. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishders, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium, and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spezifikation

Chemische Identifikatoren

10043-52-4
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
[Cl-].[Cl-].[Ca++]
CaCl2
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312

Spezifikation

>1600°C
10043-52-4
Granular Powder
CaCl2
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium;dichloride
5284359
110.99
White to gray granular powder
Calcium chloride
782°C
White to Gray
93%
MFCD00010903
Freely soluble in water,ethanol,methanol,and acetic acid. Insoluble in liquid ammonia,dimethyl sulfoxide,and ethyl acetate.
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312
Hygroscopic
3.08g/cm3
Videos
Sicherheit und Handhabung

Sicherheit und Handhabung

P264b-P280i-P305+P351+P338-P312-P501c

H319-H500

missing translation for 'einecsNumber' : 233-140-8

missing translation for 'rtecsNumber' : EV9800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Produktidentifikator
  • Calcium chloride, anhydrous

Signalwort
  • Warning

Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2

Gefahrenhinweis
  • H319-Causes serious eye irritation.

Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

RUO – Research Use Only