missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, anhydrous, ACS, 96.0% min, Thermo Scientific Chemicals

Typical ionic halide, often used as a dessicant

£46.73 - £80.00

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11443840
View Documents
Thermo Scientific Alfa Aesar
089866.18
50 g
£46.73
50g
Please to purchase this item. Need a web account? Register with us today!
11453840
View Documents
Thermo Scientific Alfa Aesar
089866.30
250 g
£80.00
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Typically an ionic halide often used as a dessicant,Calcium chloride is used as a drying and dehydrating agent for organic liquids and gases and for solids in desiccators. Common applications include brine for refrigeration plants and ice and dust control on roads. Use of calcium chloride and urea has been reported to significantly increase average fruit weight and ascorbic acid content in pomegranate. The strawberry fruit storability can also be improved by edible coating of calcium chloride. It is used in drying tubes in research laboratories, salt/chemical based dehumidifiers, fire-extinguishers, plastics, ceramic slipware, as flux in the Davy process for the manufacture of sodium and to increase water hardness in swimming pool. It provides inhibition of swelling clays in the water phase of invert emulsion drilling fluids. CaCl2-Pybox (pyridine flanked by two oxazoline groups) has been reported to be an efficient chiral catalyst for asymmetric 1,4-addition reactions of 1,3-dicarbonyl compounds with nitroalkenes, affording gamma-nitro carbonyl compounds in high enantioselectivities.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
Specifications

Chemical Identifiers

10043-52-4
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
calcium dichloride
CaCl2
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312
[Cl-].[Cl-].[Ca++]

Specifications

>1600°C
10043-52-4
96.0% min.
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
5284359
110.99
ACS
Calcium chloride, Anhydrous
782°C
Powder
CaCl2
14,1659
Freely soluble in water,ethanol,methanol and acetic acid. Insoluble in liquid ammonia,dimethyl sulfoxide and ethyl acetate.
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312
≥96.0%
Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H319
Causes serious eye irritation.

P264b-P280i-P305+P351+P338-P312-P501c

H319-H500

missing translation for 'einecsNumber' : 233-140-8

missing translation for 'rtecsNumber' : EV9800000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged under Argon

Recommended Storage : Ambient temperatures

Product Identifier
  • Calcium chloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Calcium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only