missing translation for 'onlineSavingsMsg'
Learn More
Calcium Chloride, Anhydrous, granular, Honeywell Fluka™ Product Code.: 15687720

Calcium Chloride, Anhydrous, granular, Honeywell Fluka™

Product Code. 15687720 Shop All Honeywell Fluka Products
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 15687720

Brand: Honeywell-Fluka C1016100G

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Anhydrous, granular, ≤7.0 mm,≥93.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium;dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Quantity 100 g
Linear Formula CaCl2
UN Number NONH for all modes of transport
Formula Weight 110.98g/mol
Percent Purity ≥93%
Vapor Pressure 0.01 mmHg (20°C)
Chemical Name or Material Calcium chloride
compliance-icons
Product Identifier
  • Calcium chloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Chloride, Anhydrous, granular, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.