missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride, Anhydrous, powder, ≥97%, Honeywell Fluka™

Product Code. 15667760
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Anhydrous, powder,≥97%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Boiling Point 1,900°C
Melting Point 772°C (literature)
CAS Max % 100.0000%
Physical Form Powder
Quantity 5 kg
Linear Formula CaCl2
UN Number NONH for all modes of transport
IUPAC Name calcium;dichloride
Formula Weight 110.98
Percent Purity >97%
Packaging Poly Bottle
Vapor Pressure 0.01 mmHg (20°C)
Density 2.150 g/cm3
Chemical Name or Material Calcium chloride, Anhydrous, powder, ≥97%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'