missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 11994171
2.5 kg, Plastic bottle
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2·2H2O
Molecular Weight (g/mol) 147.02
MDL Number MFCD00149613

Specifications

Melting Point 175.0°C
Color White
pH 4.5 to 8.5 (5% soln. at 25°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 kg
Assay Percent Range 99.0 to 105% (Complexometry)
Linear Formula CaCl2·2H2O
Fieser 01,104
Merck Index 15,1661
Solubility Information Solubility in water: 1000g/L (0°C). Other solubilities: freely soluble in alcohol
Formula Weight 147.02
Percent Purity ≥99%
Grade ACS Reagent
Packaging Plastic bottle
Specific Gravity 0.83
Density 0.8300g/mL
Chemical Name or Material Calcium chloride dihydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467384 Certificate TypeCertificate of Analysis Date06/11/2024 Catalog Number423520025,423525000,423520250
Lot NumberA0465217 Certificate TypeCertificate of Analysis Date05/08/2024 Catalog Number423520025,423525000,423520250
Lot NumberA0461664 Certificate TypeCertificate of Analysis Date05/04/2024 Catalog Number423520025,423525000,423520250
Lot NumberA0459989 Certificate TypeCertificate of Analysis Date12/02/2024 Catalog Number423520025,423525000,423520250
Lot NumberA0444254 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number423520025,423525000,423520250
compliance-icons
Product Identifier
  • Calcium chloride dihydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium chloride dihydrate, 99+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.